ContactUsLinkComponent
IL25 Mouse Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 143 amino acids and having a m olecular mass of 15.8kDa. The Mouse SF20 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80103-10
Supplier:  Biorbyt
IL25 Mouse Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 143 amino acids and having a m olecular mass of 15.8kDa. The Mouse SF20 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80103-10
Supplier:  Biorbyt