Noggin Human Recombinant produced in E.coli is non-glycosylated, non-disulfide-linked homodimer consisting of two 206 amino acid pol ypeptide chains, having total molecular mass of approximately 46.2 kDa (each chain 23.1 kDa).Noggin is purif 1 * 20 µG
Catalog # BIRBORB80092-20
Supplier: Biorbyt
Frequently Bought Together