ContactUsLinkComponent
MCSF Human Recombinant produced in E.coli is disulfide linked homodimer, non-glycosylated, polypeptide chain containing x 159 amino acids and having total molecular mass of 36.8 KD.MCSF is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80088-10
Supplier:  Biorbyt
MCSF Human Recombinant produced in E.coli is disulfide linked homodimer, non-glycosylated, polypeptide chain containing x 159 amino acids and having total molecular mass of 36.8 KD.MCSF is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80088-10
Supplier:  Biorbyt