ContactUsLinkComponent
Interleukin33 Mouse recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 158 amino acids and h aving a molecular mass of 17.5 kDa. 1 * 10 µG
  BIRBORB80084-10
 :  Biorbyt
Interleukin33 Mouse recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 158 amino acids and h aving a molecular mass of 17.5 kDa. 1 * 10 µG
  BIRBORB80084-10
 :  Biorbyt
Frequently Bought Together