ContactUsLinkComponent
Human IL7 Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 153 amino acids and having a mol ecular mass of 17412 Dalton. The IL7 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80066-10
Supplier:  Biorbyt
Human IL7 Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 153 amino acids and having a mol ecular mass of 17412 Dalton. The IL7 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80066-10
Supplier:  Biorbyt