ContactUsLinkComponent
IL1 beta Monkey Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 153 amino acids and having a molecu lar mass of 17.3 kDa. The IL-1b is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80053-10
Supplier:  Biorbyt
IL1 beta Monkey Recombinant produced in E.coli is non-glycosylated, Polypeptide chain containing 153 amino acids and having a molecu lar mass of 17.3 kDa. The IL-1b is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB80053-10
Supplier:  Biorbyt