ContactUsLinkComponent
Recombinant Rhesus Macaqu Granulocyte MCSF Monkey Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain co ntaining 127 amino acids and having a molecular mass of 14.4 kDa. GMCSF is purified by proprietary chromatog 1 * 2 µG
Catalog # BIRBORB80034-2
Supplier:  Biorbyt
Recombinant Rhesus Macaqu Granulocyte MCSF Monkey Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain co ntaining 127 amino acids and having a molecular mass of 14.4 kDa. GMCSF is purified by proprietary chromatog 1 * 2 µG
Catalog # BIRBORB80034-2
Supplier:  Biorbyt