ContactUsLinkComponent
Granulocyte MCSF Rat Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 128 amino acids and having a molecular mass of 14590.65 Dalton. GMCSF Rat Recombinant is purified by proprietary chromatographic 1 * 5 µG
Catalog # BIRBORB80033-5
Supplier:  Biorbyt
Granulocyte MCSF Rat Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 128 amino acids and having a molecular mass of 14590.65 Dalton. GMCSF Rat Recombinant is purified by proprietary chromatographic 1 * 5 µG
Catalog # BIRBORB80033-5
Supplier:  Biorbyt