ContactUsLinkComponent
The ELISA Sreening Kit is a competitive enzyme-labeled immunoassay for the quantitation of aflatoxins in nuts and grain products. Th e test principle is based on a direct competitive enzyme-linked immunosorbent assay (dcELISA). In the assay, 1 * 96 items
Catalog # BIRBORB59528-96
Supplier:  Biorbyt
The ELISA Sreening Kit is a competitive enzyme-labeled immunoassay for the quantitation of aflatoxins in nuts and grain products. Th e test principle is based on a direct competitive enzyme-linked immunosorbent assay (dcELISA). In the assay, 1 * 96 items
Catalog # BIRBORB59528-96
Supplier:  Biorbyt