ContactUsLinkComponent
The ELISA Sreening Kit is an immunochemical test for the quantitation of Microcystins (MCY) in algae or water samples. The test prin ciple is based on a direct competitive enzyme-linked Immunosorbent assay (dcELISA). In the assay, MCY toxin 1 * 96 items
Catalog # BIRBORB59527-96
Supplier:  Biorbyt
The ELISA Sreening Kit is an immunochemical test for the quantitation of Microcystins (MCY) in algae or water samples. The test prin ciple is based on a direct competitive enzyme-linked Immunosorbent assay (dcELISA). In the assay, MCY toxin 1 * 96 items
Catalog # BIRBORB59527-96
Supplier:  Biorbyt