ContactUsLinkComponent
Exendin-4 Recombinant is novel 39-amino acids peptide having a molecular mass of 4186.7 Dalton, which shares 53%, sequence homology with GLP-17-36 amide and interacts with the same membrane receptor. The Exendin-4 is purified by proprietary 1 * 100 µG
Catalog # BIRBORB80166-100
Supplier:  Biorbyt
Exendin-4 Recombinant is novel 39-amino acids peptide having a molecular mass of 4186.7 Dalton, which shares 53%, sequence homology with GLP-17-36 amide and interacts with the same membrane receptor. The Exendin-4 is purified by proprietary 1 * 100 µG
Catalog # BIRBORB80166-100
Supplier:  Biorbyt