ContactUsLinkComponent
MIP-3 Alpha Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 70 amino acids and havi ng a molecular mass of kDa. The MIP-3a is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80153-20
Supplier:  Biorbyt
MIP-3 Alpha Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 70 amino acids and havi ng a molecular mass of kDa. The MIP-3a is purified by proprietary chromatographic techniques. 1 * 20 µG
Catalog # BIRBORB80153-20
Supplier:  Biorbyt