ContactUsLinkComponent
Human cystatin C (or cystatin 3), which is composed of 120 amino acid residues, belongs to the cystatins superfamilly that inactivat es lysosomal cysteine proteinases. As a strongly cationic and low-molecular weight (13.4 kDa) protein, it is 1 * 96 items
Catalog # BIRBORB54808-96
Supplier:  Biorbyt
Human cystatin C (or cystatin 3), which is composed of 120 amino acid residues, belongs to the cystatins superfamilly that inactivat es lysosomal cysteine proteinases. As a strongly cationic and low-molecular weight (13.4 kDa) protein, it is 1 * 96 items
Catalog # BIRBORB54808-96
Supplier:  Biorbyt