ContactUsLinkComponent
Sandwich ELISA kit for Quantitative Detection for Mouse GDNF.Glial cell line-derived neurotrophic factor (GDNF) is a glycosylated, d isulfide-bonded homodimer that is a distantly related member of the transforming growth factor-beta superfam 1 * 96 items
Catalog # BIRBORB99235-96
Supplier:  Biorbyt
Sandwich ELISA kit for Quantitative Detection for Mouse GDNF.Glial cell line-derived neurotrophic factor (GDNF) is a glycosylated, d isulfide-bonded homodimer that is a distantly related member of the transforming growth factor-beta superfam 1 * 96 items
Catalog # BIRBORB99235-96
Supplier:  Biorbyt