ContactUsLinkComponent
HCV-N-G6a (residues 2-119) protein.The protein contains the HCV core nucleocapsid immunodominant regions, amino acids: 1-102.Hepatit is C Virus core proteins are purified by proprietarychromatographic techniques.The hepatitis C virus (HCV) c 1 * 1 mg
  BIRBORB97896-1
 :  Biorbyt
HCV-N-G6a (residues 2-119) protein.The protein contains the HCV core nucleocapsid immunodominant regions, amino acids: 1-102.Hepatit is C Virus core proteins are purified by proprietarychromatographic techniques.The hepatitis C virus (HCV) c 1 * 1 mg
  BIRBORB97896-1
 :  Biorbyt
Frequently Bought Together