ContactUsLinkComponent
Sandwich Elisa kit for Quantitative Detection for Human Galectin-3 or LGALS3, Galectin-3GAL3, also known as LGALS3, MAC2 or GALBP, i s a member of the lectin family, of which 14 mammalian galectins have been identified. Galectin-3 is encoded 1 * 96 Tests
Catalog # BIRBORB96785-96
Supplier:  Biorbyt
Sandwich Elisa kit for Quantitative Detection for Human Galectin-3 or LGALS3, Galectin-3GAL3, also known as LGALS3, MAC2 or GALBP, i s a member of the lectin family, of which 14 mammalian galectins have been identified. Galectin-3 is encoded 1 * 96 Tests
Catalog # BIRBORB96785-96
Supplier:  Biorbyt