ContactUsLinkComponent
Recombinant antigen consists of HBV immunodominant region containing 10 precore residues at its N-terminus and 1-149 residues from t he end of precore to its C-terminus. HBV e-Antigen was expressed from E.coli, the purified recombinant core 1 * 25 µG
Catalog # BIRBORB81636-25
Supplier:  Biorbyt
Recombinant antigen consists of HBV immunodominant region containing 10 precore residues at its N-terminus and 1-149 residues from t he end of precore to its C-terminus. HBV e-Antigen was expressed from E.coli, the purified recombinant core 1 * 25 µG
Catalog # BIRBORB81636-25
Supplier:  Biorbyt