ContactUsLinkComponent
PFN1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 140 amino acids (1-140 a.a.) a nd having a molecular mass of 15kDa.The PFN1 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB81270-10
Supplier:  Biorbyt
PFN1 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 140 amino acids (1-140 a.a.) a nd having a molecular mass of 15kDa.The PFN1 is purified by proprietary chromatographic techniques. 1 * 10 µG
Catalog # BIRBORB81270-10
Supplier:  Biorbyt