ContactUsLinkComponent
PDCD5 Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 125 amino acids and having a molecul ar mass of 14 kDa. 1 * 5 µG
Catalog # BIRBORB81264-5
Supplier:  Biorbyt
PDCD5 Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 125 amino acids and having a molecul ar mass of 14 kDa. 1 * 5 µG
Catalog # BIRBORB81264-5
Supplier:  Biorbyt