ContactUsLinkComponent
NUCB2 Mouse Recombinant fused to N-terminal His-Tag produced in E.coli is a single, non-glycosylated polypeptide chain containing 41 7 amino acids and having a molecular mass of 49 kDa. 1 * 25 µG
Catalog # BIRBORB81241-25
Supplier:  Biorbyt
NUCB2 Mouse Recombinant fused to N-terminal His-Tag produced in E.coli is a single, non-glycosylated polypeptide chain containing 41 7 amino acids and having a molecular mass of 49 kDa. 1 * 25 µG
Catalog # BIRBORB81241-25
Supplier:  Biorbyt