ContactUsLinkComponent
MYL2 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 186 amino acids (1-166) and ha ving a molecular mass of 20.9 kDa. MYL2 is fused to 20 amino acid His Tag fused at N-terminus and purified b 1 * 5 µG
Catalog # BIRBORB81225-5
Supplier:  Biorbyt
MYL2 Human Recombinant produced in E.coli is a single, non-glycosylated, polypeptide chain containing 186 amino acids (1-166) and ha ving a molecular mass of 20.9 kDa. MYL2 is fused to 20 amino acid His Tag fused at N-terminus and purified b 1 * 5 µG
Catalog # BIRBORB81225-5
Supplier:  Biorbyt