ContactUsLinkComponent
MSH6 Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 51 amino acids (350-400 a.a.) having total Mw of 33 kDa. Human MSH6 is fused to GST tag and purified by proprietary chromatographic techni 1 * 2 µG
Catalog # BIRBORB81223-2
Supplier:  Biorbyt
MSH6 Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 51 amino acids (350-400 a.a.) having total Mw of 33 kDa. Human MSH6 is fused to GST tag and purified by proprietary chromatographic techni 1 * 2 µG
Catalog # BIRBORB81223-2
Supplier:  Biorbyt