ContactUsLinkComponent
ANXA8 Human Recombinant produced in E.coli is signle, non-glycosylated, polypeptide chain containing 347 amino acids (1-327 a.a.)and having total molecular mass of 39 kDa. ANXA8 is fused to 20 amino acid His Tag at N-terminus and is purifie 1 * 5 µG
Catalog # BIRBORB80919-5
Supplier:  Biorbyt
ANXA8 Human Recombinant produced in E.coli is signle, non-glycosylated, polypeptide chain containing 347 amino acids (1-327 a.a.)and having total molecular mass of 39 kDa. ANXA8 is fused to 20 amino acid His Tag at N-terminus and is purifie 1 * 5 µG
Catalog # BIRBORB80919-5
Supplier:  Biorbyt