ContactUsLinkComponent
Human Angiostatin kringles 1-4 is produced from Human Fluid is glycosylated polypeptide chain which migrates as doublet 50 kDa on SD S-PAGE. The Ang K1-4 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80911-2
Supplier:  Biorbyt
Human Angiostatin kringles 1-4 is produced from Human Fluid is glycosylated polypeptide chain which migrates as doublet 50 kDa on SD S-PAGE. The Ang K1-4 is purified by proprietary chromatographic techniques. 1 * 2 µG
Catalog # BIRBORB80911-2
Supplier:  Biorbyt