ContactUsLinkComponent
CKMT1 Human Recombinant without C-terminal Lysine on both chains produced in Pichia Pastoris is glycosylated 47kDa polypeptide chain having an identical amino acid sequence compared to the native enzyme, purified under non-denaturing condit 1 * 10 µG
Catalog # BIRBORB80893-10
Supplier:  Biorbyt
CKMT1 Human Recombinant without C-terminal Lysine on both chains produced in Pichia Pastoris is glycosylated 47kDa polypeptide chain having an identical amino acid sequence compared to the native enzyme, purified under non-denaturing condit 1 * 10 µG
Catalog # BIRBORB80893-10
Supplier:  Biorbyt