ContactUsLinkComponent
Human CKM produced in Human Cardiac Tissues having a molecular mass of 82 kDa and PI 5.0-7.5. It's the skeletal muscle specific crea tine kinase and is elevated in case of strenuous exercise and in degenerative muscle disease. 1 * 1 mg
Catalog # BIRBORB80892-1
Supplier:  Biorbyt
Human CKM produced in Human Cardiac Tissues having a molecular mass of 82 kDa and PI 5.0-7.5. It's the skeletal muscle specific crea tine kinase and is elevated in case of strenuous exercise and in degenerative muscle disease. 1 * 1 mg
Catalog # BIRBORB80892-1
Supplier:  Biorbyt