ContactUsLinkComponent
CKMBITI Human Recombinant produced in Pichia Pastoris is glycosylated polypeptide chain having an identical amino acid sequence comp ared to the native enzyme, purified under non-denaturing conditions and reacts with polyclonal antibodies to 1 * 10 µG
Catalog # BIRBORB80890-10
Supplier:  Biorbyt
CKMBITI Human Recombinant produced in Pichia Pastoris is glycosylated polypeptide chain having an identical amino acid sequence comp ared to the native enzyme, purified under non-denaturing conditions and reacts with polyclonal antibodies to 1 * 10 µG
Catalog # BIRBORB80890-10
Supplier:  Biorbyt