ContactUsLinkComponent
Recombinant Listeriolysin lacking both the signal secretion sequence and the PEST- like sequence. LLO-PEST minus is composed of 471 amino acids, starting from amino acid 60 to amino acid 529, with the addition of methionine in its NH2-termi 1 * 50 µG
  BIRBORB81200-50
 :  Biorbyt
Recombinant Listeriolysin lacking both the signal secretion sequence and the PEST- like sequence. LLO-PEST minus is composed of 471 amino acids, starting from amino acid 60 to amino acid 529, with the addition of methionine in its NH2-termi 1 * 50 µG
  BIRBORB81200-50
 :  Biorbyt
Frequently Bought Together