ContactUsLinkComponent
Recombinant Chimeric Chagas Multiantigen (MACH) (Trypanosoma cruzi) produced in E.coli, containing 6-His tag and having an Mw of 9.9 kDa. 1 * 5 µG
Catalog # BIRBORB81539-5
Supplier:  Biorbyt
Recombinant Chimeric Chagas Multiantigen (MACH) (Trypanosoma cruzi) produced in E.coli, containing 6-His tag and having an Mw of 9.9 kDa. 1 * 5 µG
Catalog # BIRBORB81539-5
Supplier:  Biorbyt