ContactUsLinkComponent
HIF1A Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 231 amino acids (576-785 a.a.) and having a molecular mass of 25.1 kDa. The HIF1A is fused to 21 amino acid His-Tag at N-terminus and puri 1 * 10 µG
Catalog # BIRBORB81158-10
Supplier:  Biorbyt
HIF1A Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 231 amino acids (576-785 a.a.) and having a molecular mass of 25.1 kDa. The HIF1A is fused to 21 amino acid His-Tag at N-terminus and puri 1 * 10 µG
Catalog # BIRBORB81158-10
Supplier:  Biorbyt