ContactUsLinkComponent
Recombinant Full-Length H1N1 A/Solomon Islands/03/2006 is glycosylated with N-linked sugars, produced using baculovirus vectors in i nsect cells. 1 * 10 µG
Catalog # BIRBORB81777-10
Supplier:  Biorbyt
Recombinant Full-Length H1N1 A/Solomon Islands/03/2006 is glycosylated with N-linked sugars, produced using baculovirus vectors in i nsect cells. 1 * 10 µG
Catalog # BIRBORB81777-10
Supplier:  Biorbyt