ContactUsLinkComponent
Recombinant Full-Length H1N1 New Caledonia/20/99 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect cells and its Mw is approximately 72 kDa.The insect cells infected with A9440.1a recombinant baculovirus exp 1 * 10 µG
Catalog # BIRBORB81776-10
Supplier:  Biorbyt
Recombinant Full-Length H1N1 New Caledonia/20/99 is glycosylated with N-linked sugars, produced using baculovirus vectors in insect cells and its Mw is approximately 72 kDa.The insect cells infected with A9440.1a recombinant baculovirus exp 1 * 10 µG
Catalog # BIRBORB81776-10
Supplier:  Biorbyt