ContactUsLinkComponent
Geminin Human Recombinant fused to N-terminal His-Tag produced in E.coli is a single, non-glycosylated polypeptide chain containing 245 amino acids and having a molecular mass of 27.7 kDa. 1 * 20 µG
Catalog # BIRBORB81144-20
Supplier:  Biorbyt
Geminin Human Recombinant fused to N-terminal His-Tag produced in E.coli is a single, non-glycosylated polypeptide chain containing 245 amino acids and having a molecular mass of 27.7 kDa. 1 * 20 µG
Catalog # BIRBORB81144-20
Supplier:  Biorbyt