ContactUsLinkComponent
GLIPR2 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 174 amino acids (1-154) havin g a molecular mass of 19.3 kDa.The GLIPR2 is fused to 20 amino acid His-Tag at N-terminus and purified by st 1 * 5 µG
Catalog # BIRBORB81143-5
Supplier:  Biorbyt
GLIPR2 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 174 amino acids (1-154) havin g a molecular mass of 19.3 kDa.The GLIPR2 is fused to 20 amino acid His-Tag at N-terminus and purified by st 1 * 5 µG
Catalog # BIRBORB81143-5
Supplier:  Biorbyt