ContactUsLinkComponent
Ferric Uptake Regulator Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 148 amino acids an d having a molecular mass of 16 kDa. 1 * 25 µG
Catalog # BIRBORB81128-25
Supplier:  Biorbyt
Ferric Uptake Regulator Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 148 amino acids an d having a molecular mass of 16 kDa. 1 * 25 µG
Catalog # BIRBORB81128-25
Supplier:  Biorbyt