ContactUsLinkComponent
FSCN1 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 513 amino acids (1-493 a.a.) a nd having a molecular mass of 56.7 kDa. The FSCN1 is fused to 20 amino acid His-Tag at N-terminus and purifi 1 * 5 µG
Catalog # BIRBORB81127-5
Supplier:  Biorbyt
FSCN1 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 513 amino acids (1-493 a.a.) a nd having a molecular mass of 56.7 kDa. The FSCN1 is fused to 20 amino acid His-Tag at N-terminus and purifi 1 * 5 µG
Catalog # BIRBORB81127-5
Supplier:  Biorbyt