ContactUsLinkComponent
FABP7 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 132 amino acids and having a m olecular mass of 19.39kDa. FABP7 is fused to His-Tag at N-terminus and purified by standard chromatography t 1 * 5 µG
Catalog # BIRBORB81107-5
Supplier:  Biorbyt
FABP7 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 132 amino acids and having a m olecular mass of 19.39kDa. FABP7 is fused to His-Tag at N-terminus and purified by standard chromatography t 1 * 5 µG
Catalog # BIRBORB81107-5
Supplier:  Biorbyt