ContactUsLinkComponent
Recombinant Human Trypsin-2 is free from any animal and human sources. Recombinant Human Trypsin-2 expressed in E.coli having Mw of 24kDa is purified by standard chromatography techniques. Recombinant Human Trypsin-2 is free from foreign en 1 * 5 mg
Catalog # BIRBORB81439-5
Dodavatel:  Biorbyt
Recombinant Human Trypsin-2 is free from any animal and human sources. Recombinant Human Trypsin-2 expressed in E.coli having Mw of 24kDa is purified by standard chromatography techniques. Recombinant Human Trypsin-2 is free from foreign en 1 * 5 mg
Catalog # BIRBORB81439-5
Dodavatel:  Biorbyt
Související informace
Frequently Bought Together