ContactUsLinkComponent
TREM1 Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 180 amino acids fragment (21- 200) corresponding to mature soluble extracellular domain fragment of TREM-1, having total molecular mass of 1 * 10 µG
Catalog # BIRBORB81430-10
Supplier:  Biorbyt
TREM1 Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 180 amino acids fragment (21- 200) corresponding to mature soluble extracellular domain fragment of TREM-1, having total molecular mass of 1 * 10 µG
Catalog # BIRBORB81430-10
Supplier:  Biorbyt