ContactUsLinkComponent
Eledoisin's molecular weight is 1188.4 having an amino acid sequence of Glp-Pro-Ser-Lys-Asp-Ala-Phe-Ile-Gly-Leu-Met-NH2 and a molecu lar formula of C54H85N13O15S. 1 * 100 mg
  BIRBORB81085-100
 :  Biorbyt
Eledoisin's molecular weight is 1188.4 having an amino acid sequence of Glp-Pro-Ser-Lys-Asp-Ala-Phe-Ile-Gly-Leu-Met-NH2 and a molecu lar formula of C54H85N13O15S. 1 * 100 mg
  BIRBORB81085-100
 :  Biorbyt
Frequently Bought Together