SOX9 Human Recombinant produced in E.coli is a single, non-glycosylated, Polypeptide chain containing 150 amino acids (1-150 a.a.) f used to GST tag and having total molecular mass of 43kDa. The SOX9 is purified by proprietary chromatographi 1 * 10 µG
Katalogové číslo BIRBORB81380-10
Supplier: Biorbyt
Frequently Bought Together