ContactUsLinkComponent
Recombinant C.elegans SNAP-25 produced in E.coli is a single, non-glycosylated polypeptide chain containing 207 amino acids and havi ng a molecular mass of 23kDa. SNAP-25 gene was amplified by PCR from C.elegans and cloned into an E.coli exp 1 * 50 µG
Catalog # BIRBORB81371-50
Supplier:  Biorbyt
Recombinant C.elegans SNAP-25 produced in E.coli is a single, non-glycosylated polypeptide chain containing 207 amino acids and havi ng a molecular mass of 23kDa. SNAP-25 gene was amplified by PCR from C.elegans and cloned into an E.coli exp 1 * 50 µG
Catalog # BIRBORB81371-50
Supplier:  Biorbyt