ContactUsLinkComponent
SMAD4 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 572 amino acids (1-552) and ha ving a molecular mass of 62.6 kDa. SMAD4 is fused to 20 amino acid His-Tag at N-Terminus and purified by sta 1 * 10 µG
Catalog # BIRBORB81368-10
Supplier:  Biorbyt
SMAD4 Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 572 amino acids (1-552) and ha ving a molecular mass of 62.6 kDa. SMAD4 is fused to 20 amino acid His-Tag at N-Terminus and purified by sta 1 * 10 µG
Catalog # BIRBORB81368-10
Supplier:  Biorbyt