ContactUsLinkComponent
HIV-1 TAT Recombinant- produced in E.coli is a single, non-glycosylated, polypeptide chain containing 100 amino acids encoded by two exons and having chain having a molecular mass of 21 kDa. 1 * 2 µG
Catalog # BIRBORB81724-2
Supplier:  Biorbyt
HIV-1 TAT Recombinant- produced in E.coli is a single, non-glycosylated, polypeptide chain containing 100 amino acids encoded by two exons and having chain having a molecular mass of 21 kDa. 1 * 2 µG
Catalog # BIRBORB81724-2
Supplier:  Biorbyt