ContactUsLinkComponent
Rabbit Skeletal Muscle Troponin-C is a single, glycosylated, polypeptide chain having a molecular mass of 18,096 Dalton. Troponin-C is one of three subunits that form the Troponin complex of striated muscle thin filaments. Skeletal muscle T 1 * 10 mg
Catalog # BIRBORB81328-10
Dodavatel:  Biorbyt
Rabbit Skeletal Muscle Troponin-C is a single, glycosylated, polypeptide chain having a molecular mass of 18,096 Dalton. Troponin-C is one of three subunits that form the Troponin complex of striated muscle thin filaments. Skeletal muscle T 1 * 10 mg
Catalog # BIRBORB81328-10
Dodavatel:  Biorbyt
Související informace
Frequently Bought Together