ContactUsLinkComponent
RND3 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 261 amino acids (1-241 a.a.) an d having a molecular mass of 29.2 kDa. The RND3 is fused to 20 amino acid His-Tag at N-terminus and purified 1 * 5 µG
Catalog # BIRBORB81322-5
Supplier:  Biorbyt
RND3 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 261 amino acids (1-241 a.a.) an d having a molecular mass of 29.2 kDa. The RND3 is fused to 20 amino acid His-Tag at N-terminus and purified 1 * 5 µG
Catalog # BIRBORB81322-5
Supplier:  Biorbyt