ContactUsLinkComponent
HSA Human Recombinant produced in Plant is non-glycosylated, polypeptide chain containing 585 amino acids and having a molecular mas s of 67 kDa.The optimum concentration for recombinant Albumin to be used in cell culture ranges between 0.5g 1 * 10 mg
Catalog # BIRBORB81321-10
Supplier:  Biorbyt
HSA Human Recombinant produced in Plant is non-glycosylated, polypeptide chain containing 585 amino acids and having a molecular mas s of 67 kDa.The optimum concentration for recombinant Albumin to be used in cell culture ranges between 0.5g 1 * 10 mg
Catalog # BIRBORB81321-10
Supplier:  Biorbyt