ContactUsLinkComponent
RHEB Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 197 amino acids (1-181 amino ac ids) and having a molecular mass of 21.7 kDa.The RHEB is fused to T7-tag at N-terminus (16 a.a.) and is puri 1 * 10 µG
Catalog # BIRBORB81318-10
Supplier:  Biorbyt
RHEB Human Recombinant produced in E.coli is a single, non-glycosylated polypeptide chain containing 197 amino acids (1-181 amino ac ids) and having a molecular mass of 21.7 kDa.The RHEB is fused to T7-tag at N-terminus (16 a.a.) and is puri 1 * 10 µG
Catalog # BIRBORB81318-10
Supplier:  Biorbyt