ContactUsLinkComponent
CBFB Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 182 amino acids (1-202 a.a.) an d having a molecular mass of 23.6 kDa. The CBFB is fused to 20 amino acid His-Tag at N-terminus and purified 1 * 2 µG
Catalog # BIRBORB80999-2
Supplier:  Biorbyt
CBFB Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 182 amino acids (1-202 a.a.) an d having a molecular mass of 23.6 kDa. The CBFB is fused to 20 amino acid His-Tag at N-terminus and purified 1 * 2 µG
Catalog # BIRBORB80999-2
Supplier:  Biorbyt