ContactUsLinkComponent
RCN1 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 341 amino acids (30-331 a.a.) a nd having a molecular mass of 22.5 kDa. The RCN1 is fused to 39 amino acid His-Tag at N-terminus and purifie 1 * 10 µG
Catalog # BIRBORB81310-10
Supplier:  Biorbyt
RCN1 Recombinant Human produced in E.coli is a single, non-glycosylated polypeptide chain containing 341 amino acids (30-331 a.a.) a nd having a molecular mass of 22.5 kDa. The RCN1 is fused to 39 amino acid His-Tag at N-terminus and purifie 1 * 10 µG
Catalog # BIRBORB81310-10
Supplier:  Biorbyt